Anti-FOS Chicken polyclonal antibody

ABCAAB14285-50EA 619 EUR
ABCAAB14285-50
Anti-FOS Chicken polyclonal antibody
Type: Primary
Antigen: FOS (FBJ murine osteosarcoma viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR