Anti-GNAI3 Chicken polyclonal antibody unconjugated

Proveedor: Abcam
ABCAAB14246-50EA 0 EUR
ABCAAB14246-50
Anti-GNAI3 Chicken polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: GNAI3 (guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR