Anti-CDC25C Chicken polyclonal antibody

ABCAAB14240-50EA 588 EUR
ABCAAB14240-50
Anti-CDC25C Chicken polyclonal antibody
Type: Primary
Antigen: CDC25C (cell division cycle 25C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR