Anti-UTS2 Rabbit polyclonal antibody

ABCAAB14200-50EA 604 EUR
ABCAAB14200-50
Anti-UTS2 Rabbit polyclonal antibody
Type: Primary
Antigen: UTS2 (urotensin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR