Anti-GAST Rabbit polyclonal antibody unconjugated

Proveedor: Abcam
ABCAAB14182-50EA 781 EUR
ABCAAB14182-50
Anti-GAST Rabbit polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: GAST (gastrin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR