Anti-PTH Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB14164-50EA 739 EUR
ABCAAB14164-50
Anti-PTH Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PTH (parathyroid hormone)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR