Anti-CSF2 Chicken polyclonal antibody unconjugated

Proveedor: Abcam
ABCAAB14024-50EA 0 EUR
ABCAAB14024-50
Anti-CSF2 Chicken polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: CSF2 (colony stimulating factor 2 (granulocyte-macrophage))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR