Anti-CDH1 Chicken polyclonal antibody

ABCAAB14015-50EA 653 EUR
ABCAAB14015-50
Anti-CDH1 Chicken polyclonal antibody
Type: Primary
Antigen: CDH1 (cadherin 1, type 1, E-cadherin (epithelial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR