Anti-ATP7A Chicken polyclonal antibody unconjugated

Proveedor: Abcam
ABCAAB13995-50EA 0 EUR
ABCAAB13995-50
Anti-ATP7A Chicken polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: ATP7A (ATPase, Cu++ transporting, alpha polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR