Anti-ADRB2 Chicken Polyclonal Antibody

Proveedor: Abcam
ABCAAB13989-50EA 779 EUR
ABCAAB13989-50
Anti-ADRB2 Chicken Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ADRB2 (adrenoceptor beta 2, surface)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR