Anti-ATF2 Mouse monoclonal antibody [clone: 103C411]

ABCAAB13921-50EA 577 EUR
ABCAAB13921-50
Anti-ATF2 Mouse monoclonal antibody [clone: 103C411]
Type: Primary
Antigen: ATF2 (activating transcription factor 2)
Clonality: monoclonal
Clone: 103C411
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR