Anti-Hepatitis C Virus NS4A Mouse monoclonal antibody [clone: H24]

ABCAAB13831-100EA 604 EUR
ABCAAB13831-100
Anti-Hepatitis C Virus NS4A Mouse monoclonal antibody [clone: H24]
Type: Primary
Antigen: Hepatitis C Virus NS4A
Clonality: monoclonal
Clone: H24
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR