Anti-Hepatitis C Virus Core Antigen Mouse Monoclonal Antibody [clone: H21]

Proveedor: Abcam
ABCAAB13829-100EA 739 EUR
ABCAAB13829-100
Anti-Hepatitis C Virus Core Antigen Mouse Monoclonal Antibody [clone: H21]
Anticuerpos
Type: Primary
Antigen: Hepatitis C Virus Core Antigen
Clonality: monoclonal
Clone: H21
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR