Anti-AURKA Mouse Monoclonal Antibody [clone: 35C1]

Proveedor: Abcam
ABCAAB13824-100EA 834 EUR
ABCAAB13824-100
Anti-AURKA Mouse Monoclonal Antibody [clone: 35C1]
Anticuerpos
Type: Primary
Antigen: AURKA (aurora kinase A)
Clonality: monoclonal
Clone: 35C1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR