Anti-CCR10 Rabbit polyclonal antibody

ABCAAB13794-100EA 651 EUR
ABCAAB13794-100
Anti-CCR10 Rabbit polyclonal antibody
Type: Primary
Antigen: CCR10 (chemokine (C-C motif) receptor 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR