Anti-IL1RA antagonist Mouse monoclonal antibody [clone: 1.384]

ABCAAB13776-100EA 577 EUR
ABCAAB13776-100
Anti-IL1RA antagonist Mouse monoclonal antibody [clone: 1.384]
Type: Primary
Antigen: IL1RA antagonist
Clonality: monoclonal
Clone: 1.384
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR