Anti-SETDB2 Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB13712-100EA 787 EUR
ABCAAB13712-100
Anti-SETDB2 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SETDB2 (SET domain, bifurcated 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR