Anti-EPAS1 Rabbit polyclonal antibody

ABCAAB13704-200EA 588 EUR
ABCAAB13704-200
Anti-EPAS1 Rabbit polyclonal antibody
Type: Primary
Antigen: EPAS1 (endothelial PAS domain protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR