Anti-APEX2 Rabbit polyclonal antibody

ABCAAB13691-200EA 651 EUR
ABCAAB13691-200
Anti-APEX2 Rabbit polyclonal antibody
Type: Primary
Antigen: APEX2 (APEX nuclease (apurinic/apyrimidinic endonuclease) 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR