Anti-HTRA2 Mouse monoclonal antibody [clone: 196C429]

ABCAAB13678-100EA 651 EUR
ABCAAB13678-100
Anti-HTRA2 Mouse monoclonal antibody [clone: 196C429]
Type: Primary
Antigen: HTRA2 (HtrA serine peptidase 2)
Clonality: monoclonal
Clone: 196C429
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR