Anti-UBE2N Rabbit polyclonal antibody

ABCAAB13663-100EA 577 EUR
ABCAAB13663-100
Anti-UBE2N Rabbit polyclonal antibody
Type: Primary
Antigen: UBE2N (ubiquitin-conjugating enzyme E2N)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR