Anti-TRIM37 Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB13619-100EA 486 EUR
ABCAAB13619-100
Anti-TRIM37 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TRIM37 (tripartite motif containing 37)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR