Anti-MECP2 Rabbit polyclonal antibody

ABCAAB13607-100EA 577 EUR
ABCAAB13607-100
Anti-MECP2 Rabbit polyclonal antibody
Type: Primary
Antigen: MECP2 (methyl CpG binding protein 2 (Rett syndrome))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR