Anti-Silencer of Death Domain Rabbit polyclonal antibody

ABCAAB13595-100EA 0 EUR
ABCAAB13595-100
Anti-Silencer of Death Domain Rabbit polyclonal antibody
Type: Primary
Antigen: Silencer of Death Domain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR