Anti-NCL Mouse Monoclonal Antibody [clone: 4E2]

Proveedor: Abcam
ABCAAB13541-50EA 798 EUR
ABCAAB13541-50
Anti-NCL Mouse Monoclonal Antibody [clone: 4E2]
Anticuerpos
Type: Primary
Antigen: NCL (nucleolin)
Clonality: monoclonal
Clone: 400
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR