Anti-TAP1 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB13516-50EA 739 EUR
ABCAAB13516-50
Anti-TAP1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TAP1 (transporter 1, ATP-binding cassette, sub-family B (MDR/TAP))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR