Anti-Cytochrome P450 Reductase Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB13513-50EA 480 EUR
ABCAAB13513-50
Anti-Cytochrome P450 Reductase Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Cytochrome P450 Reductase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR