Anti-PECAM1 Mouse monoclonal antibody FITC (Fluorescein) [clone: WM59]

ABCAAB13466-100EA 257 EUR
ABCAAB13466-100
Anti-PECAM1 Mouse monoclonal antibody FITC (Fluorescein) [clone: WM59]
Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: monoclonal
Clone: WM59
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR