Anti-Laminin Mouse monoclonal antibody [clone: LN26-7]

ABCAAB13414-50EA 577 EUR
ABCAAB13414-50
Anti-Laminin Mouse monoclonal antibody [clone: LN26-7]
Type: Primary
Antigen: Laminin
Clonality: monoclonal
Clone: LN26-7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR