Anti-VTN Mouse Monoclonal Antibody [clone: VN58-1]

Proveedor: Abcam
ABCAAB13413-100EA 763 EUR
ABCAAB13413-100
Anti-VTN Mouse Monoclonal Antibody [clone: VN58-1]
Anticuerpos
Type: Primary
Antigen: VTN (vitronectin)
Clonality: monoclonal
Clone: VN58-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR