Anti-ATM Goat polyclonal antibody

ABCAAB13409-50EA 611 EUR
ABCAAB13409-50
Anti-ATM Goat polyclonal antibody
Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR