Anti-Syntaxin 1a Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB13262-50EA 753 EUR
ABCAAB13262-50
Anti-Syntaxin 1a Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Syntaxin 1a
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR