Anti-DNM1 Mouse Monoclonal Antibody [clone: D5]

Proveedor: Abcam
ABCAAB13251-50EA 0 EUR
ABCAAB13251-50
Anti-DNM1 Mouse Monoclonal Antibody [clone: D5]
Anticuerpos
Type: Primary
Antigen: DNM1 (dynamin 1)
Clonality: monoclonal
Clone: D5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR