Anti-RPS6 Rabbit polyclonal antibody

Proveedor: Abcam
ABCAAB12865-50EA 707 EUR
ABCAAB12865-50
Anti-RPS6 Rabbit polyclonal antibody
Type: Primary
Antigen: RPS6 (ribosomal protein S6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR