Anti-STX6 Mouse Monoclonal Antibody [clone: 3D10]

Proveedor: Abcam
ABCAAB12370-100EA 0 EUR
ABCAAB12370-100
Anti-STX6 Mouse Monoclonal Antibody [clone: 3D10]
Anticuerpos
Type: Primary
Antigen: STX6 (syntaxin 6)
Clonality: monoclonal
Clone: 3D10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR