Anti-KDEL Mouse Monoclonal Antibody [clone: 10C3]

Proveedor: Abcam
ABCAAB12223-100EA 798 EUR
ABCAAB12223-100
Anti-KDEL Mouse Monoclonal Antibody [clone: 10C3]
Anticuerpos
Type: Primary
Antigen: KDEL
Clonality: monoclonal
Clone: 10C3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR