Anti-HIST3H3 Mouse Monoclonal Antibody [clone: mAbcam12209]

Proveedor: Abcam
ABCAAB12209-100EA 510 EUR
ABCAAB12209-100
Anti-HIST3H3 Mouse Monoclonal Antibody [clone: mAbcam12209]
Anticuerpos
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: monoclonal
Clone: mAbcam12209
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR