Anti-TBK1 Mouse Monoclonal Antibody [clone: 108A429]

Proveedor: Abcam
ABCAAB12116-50EA 0 EUR
ABCAAB12116-50
Anti-TBK1 Mouse Monoclonal Antibody [clone: 108A429]
Anticuerpos
Type: Primary
Antigen: TBK1 (TANK-binding kinase 1)
Clonality: monoclonal
Clone: 108A429
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR