Anti-E2F6 Mouse Monoclonal Antibody [clone: TFE61]

Proveedor: Abcam
ABCAAB11952-50EA 0 EUR
ABCAAB11952-50
Anti-E2F6 Mouse Monoclonal Antibody [clone: TFE61]
Anticuerpos
Type: Primary
Antigen: E2F6 (E2F transcription factor 6)
Clonality: monoclonal
Clone: TFE61
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR