Anti-CCNA2 Mouse Monoclonal Antibody [clone: C160]

ABCAAB11925-50EA 588 EUR
ABCAAB11925-50
Anti-CCNA2 Mouse Monoclonal Antibody [clone: C160]
Type: Primary
Antigen: CCNA2 (cyclin A2)
Clonality: monoclonal
Clone: C160
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR