Anti-MYC Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB11917-50EA 756 EUR
ABCAAB11917-50
Anti-MYC Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR