Anti-JUN Rabbit Polyclonal Antibody

ABCAAB11914-500EA 588 EUR
ABCAAB11914-500
Anti-JUN Rabbit Polyclonal Antibody
Type: Primary
Antigen: JUN (jun proto-oncogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR