Anti-SMARCE1 Rabbit Polyclonal Antibody

ABCAAB11910-50EA 588 EUR
ABCAAB11910-50
Anti-SMARCE1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: SMARCE1 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR