Anti-CD180 Mouse Monoclonal Antibody [clone: MHR73]

Proveedor: Abcam
ABCAAB11880-50EA 739 EUR
ABCAAB11880-50
Anti-CD180 Mouse Monoclonal Antibody [clone: MHR73]
Anticuerpos
Type: Primary
Antigen: CD180 (CD180 molecule)
Clonality: monoclonal
Clone: MHR73
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR