Anti-C5 / C5b Mouse Monoclonal Antibody [clone: 568]

Proveedor: Abcam
ABCAAB11879-50EA 0 EUR
ABCAAB11879-50
Anti-C5 / C5b Mouse Monoclonal Antibody [clone: 568]
Anticuerpos
Type: Primary
Antigen: C5 / C5b
Clonality: monoclonal
Clone: 568
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR