Anti-MD1 Rat Monoclonal Antibody [clone: MD113]

Proveedor: Abcam
ABCAAB11860-50EA 0 EUR
ABCAAB11860-50
Anti-MD1 Rat Monoclonal Antibody [clone: MD113]
Anticuerpos
Type: Primary
Antigen: MD1
Clonality: monoclonal
Clone: MD113
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR