Anti-SFPQ Mouse Monoclonal Antibody [clone: B92]

Proveedor: Abcam
ABCAAB11825-50EA 575 EUR
ABCAAB11825-50
Anti-SFPQ Mouse Monoclonal Antibody [clone: B92]
Anticuerpos
Type: Primary
Antigen: SFPQ (splicing factor proline/glutamine-rich)
Clonality: monoclonal
Clone: B92
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR