Anti-alpha 1 Spectrin Mouse Monoclonal Antibody [clone: 17C7]

Proveedor: Abcam
ABCAAB11751-100EA 0 EUR
ABCAAB11751-100
Anti-alpha 1 Spectrin Mouse Monoclonal Antibody [clone: 17C7]
Anticuerpos
Type: Primary
Antigen: alpha 1 Spectrin
Clonality: monoclonal
Clone: 17C7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR