Anti-APOH Mouse Monoclonal Antibody [clone: 1D2]

ABCAAB11733-1EA 588 EUR
ABCAAB11733-1
Anti-APOH Mouse Monoclonal Antibody [clone: 1D2]
Type: Primary
Antigen: APOH (apolipoprotein H (beta-2-glycoprotein I))
Clonality: monoclonal
Clone: 1D2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR