Anti-KSR1 Rat Monoclonal Antibody [clone: MINT/1/4]

ABCAAB11719-50EA 604 EUR
ABCAAB11719-50
Anti-KSR1 Rat Monoclonal Antibody [clone: MINT/1/4]
Type: Primary
Antigen: KSR1 (kinase suppressor of ras 1)
Clonality: monoclonal
Clone: MINT/1/4
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR